Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106442.1 | NP_001099912.1 | 4 - 5 | 443 | 125 |
GenBank mRNA | BC168754.1 | - | 4 - 5 | 415 | 125 |
FQ213377.1 | - | 4 - 5 | 407 | 125 | |
FQ221799.1 | - | 4 - 5 | 381 | 125 | |
FQ226826.1 | - | 4 - 5 | 381 | 125 | |
FQ229540.1 | - | 4 - 5 | 415 | 125 | |
FQ232929.1 | - | 4 - 5 | 399 | 125 | |
FQ235190.1 | - | 4 - 5 | 399 | 125 |