Assay Design



Gene Transcripts

Gene Symbol
Slc15a5
Entrez Gene ID
502918
Gene Name
solute carrier family 15, member 5
Gene Aliases
RGD1564925
Chromosome Location
Chr.4: 171970620 - 172063391 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192015.1 NP_001178944.1 7 - 8 1465 89

View Full Product Details