Assay Design



Gene Transcripts

Gene Symbol
Far2
Entrez Gene ID
500368
Gene Name
fatty acyl CoA reductase 2
Gene Aliases
Mlstd1, RGD1565966
Chromosome Location
Chr.4: 182446406 - 182563472 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006237697.2 XP_006237759.1 2 - 3 1309 72
XM_575726.6 XP_575726.2 2 - 3 606 72

View Full Product Details