Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108651.1 | NP_001102121.1 | 1 - 2 | 184 | 62 |
XM_006237529.1 | XP_006237591.1 | 1 - 2 | 193 | 62 | |
GenBank mRNA | BC168221.1 | - | 1 - 2 | 142 | 62 |
FQ220508.1 | - | 1 - 2 | 184 | 62 | |
FQ229464.1 | - | 1 - 2 | 184 | 62 | |
FQ233110.1 | - | 1 - 2 | 153 | 62 | |
FQ235087.1 | - | 1 - 2 | 153 | 62 |