Assay Design



Gene Transcripts

Gene Symbol
Sec14l1
Entrez Gene ID
360668
Gene Name
SEC14-like 1 (S. cerevisiae)
Gene Aliases
RGD1563123
Chromosome Location
Chr.10: 106065712 - 106112857 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108309.1 NP_001101779.1 15 - 16 2338 103
GenBank mRNA FQ212255.1 - 15 - 16 2248 103

View Full Product Details