Assay Design



Gene Transcripts

Gene Symbol
Txndc17
Entrez Gene ID
287474
Gene Name
thioredoxin domain containing 17
Gene Aliases
Txnl5
Chromosome Location
Chr.10: 58776792 - 58779764 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105805.1 NP_001099275.1 1 - 2 224 112
GenBank mRNA BC159408.1 - 1 - 2 152 112

View Full Product Details