Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011890.1 | NP_001011890.1 | 4 - 5 | 591 | 83 |
XM_006245931.2 | XP_006245993.1 | 4 - 5 | 1035 | 83 | |
XM_006245932.2 | XP_006245994.1 | 5 - 6 | 734 | 83 | |
XM_006245933.2 | XP_006245995.1 | 3 - 4 | 530 | 83 | |
GenBank mRNA | BC079014.1 | - | 4 - 5 | 591 | 83 |