Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011890.1 | NP_001011890.1 | 6 - 7 | 847 | 73 |
XM_006245931.2 | XP_006245993.1 | 6 - 7 | 1291 | 73 | |
XM_006245932.2 | XP_006245994.1 | 7 - 8 | 990 | 73 | |
XM_006245933.2 | XP_006245995.1 | 5 - 6 | 786 | 73 | |
GenBank mRNA | BC079014.1 | - | 6 - 7 | 847 | 73 |