Assay Design



Gene Transcripts

Gene Symbol
Snf8
Entrez Gene ID
287645
Gene Name
SNF8, ESCRT-II complex subunit
Gene Aliases
D11moh34, RGD1310144
Chromosome Location
Chr.10: 83856331 - 83868621 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007804.1 NP_001007805.1 7 - 8 636 105
XM_006247185.1 XP_006247247.1 7 - 8 769 105
GenBank mRNA BC086364.1 - 7 - 8 679 105

View Full Product Details