Assay Design



Gene Transcripts

Gene Symbol
Cnot8
Entrez Gene ID
363603
Gene Name
CCR4-NOT transcription complex, subunit 8
Gene Aliases
-
Chromosome Location
Chr.10: 43538155 - 43551271 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008382.1 NP_001008383.1 4 - 5 534 68
XM_006246392.2 XP_006246454.1 3 - 4 593 68
GenBank mRNA BC085856.1 - 4 - 5 534 68

View Full Product Details