Assay Design



Gene Transcripts

Gene Symbol
Alox12
Entrez Gene ID
287454
Gene Name
arachidonate 12-lipoxygenase
Gene Aliases
12-LO, 12S-LOX
Chromosome Location
Chr.10: 56851734 - 56864047 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105798.1 NP_001099268.1 4 - 5 592 71
XM_006246659.2 XP_006246721.1 4 - 5 634 71
GenBank mRNA BC168184.1 - 4 - 5 592 71

View Full Product Details