Assay Design



Gene Transcripts

Gene Symbol
Alox12
Entrez Gene ID
287454
Gene Name
arachidonate 12-lipoxygenase
Gene Aliases
12-LO, 12S-LOX
Chromosome Location
Chr.10: 56851734 - 56864047 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105798.1 NP_001099268.1 8 - 9 1210 79
GenBank mRNA BC168184.1 - 8 - 9 1210 79

View Full Product Details