Assay Design



Gene Transcripts

Gene Symbol
Srsf2
Entrez Gene ID
494445
Gene Name
serine/arginine-rich splicing factor 2
Gene Aliases
Sfrs2
Chromosome Location
Chr.10: 105792779 - 105795985 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009720.2 NP_001009720.1 2 - 3 832 132
GenBank mRNA BC058508.1 - 2 - 3 836 132

View Full Product Details