Assay Design



Gene Transcripts

Gene Symbol
Otop3
Entrez Gene ID
287821
Gene Name
otopetrin 3
Gene Aliases
-
Chromosome Location
Chr.10: 103887045 - 103898331 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105852.1 NP_001099322.1 3 - 4 649 78

View Full Product Details