Assay Design



Gene Transcripts

Gene Symbol
Eif4a3
Entrez Gene ID
688288
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
eIF4A-III
Chromosome Location
Chr.10: 108415201 - 108425195 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100158.2 NP_001093628.1 6 - 7 652 83
GenBank mRNA BC105875.1 - 6 - 7 597 83
FQ234161.1 - 5 - 6 373 83

View Full Product Details