Assay Design
Gene Transcripts
- Gene Symbol
- Zmat3
- Entrez Gene ID
-
64394
- Gene Name
- zinc finger, matrin type 3
- Gene Aliases
-
PAG608, Wig1
- Chromosome Location
-
Chr.2: 118715229 - 118746109 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_022548.1 |
NP_071993.1 |
5 - 6 |
881 |
109 |
GenBank mRNA |
Y13148.1 |
- |
5 - 6 |
881 |
109 |
View Full Product Details