Assay Design



Gene Transcripts

Gene Symbol
Zdhhc9
Entrez Gene ID
302808
Gene Name
zinc finger, DHHC-type containing 9
Gene Aliases
-
Chromosome Location
Chr.X: 135005171 - 135040996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039016.2 NP_001034105.2 1 - 2 172 59
GenBank mRNA AY886526.1 - 1 - 2 142 59
BC129070.1 - 1 - 2 172 59

View Full Product Details