Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039016.2 | NP_001034105.2 | 3 - 4 | 615 | 126 |
XM_006257521.2 | XP_006257583.1 | 3 - 4 | 717 | 126 | |
GenBank mRNA | AY886526.1 | - | 3 - 4 | 585 | 126 |
BC129070.1 | - | 3 - 4 | 615 | 126 |