Assay Design



Gene Transcripts

Gene Symbol
Slco1c1
Entrez Gene ID
84511
Gene Name
solute carrier organic anion transporter family, member 1c1
Gene Aliases
Bsat1, Oatp14, Slc21a14
Chromosome Location
Chr.4: 175729698 - 175776749 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053441.1 NP_445893.1 3 - 4 648 78
XM_006237602.1 XP_006237664.1 4 - 5 796 78
GenBank mRNA AF306546.1 - 3 - 4 648 78

View Full Product Details