Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108653.1 | NP_001102123.1 | 1 - 2 | 642 | 90 |
XM_006237703.2 | XP_006237765.1 | 1 - 2 | 622 | 90 | |
XM_006237705.2 | XP_006237767.1 | 1 - 2 | 622 | 90 | |
XM_006237706.2 | XP_006237768.1 | 1 - 2 | 622 | 90 |