Assay Design



Gene Transcripts

Gene Symbol
Bicd1
Entrez Gene ID
362466
Gene Name
bicaudal D homolog 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.4: 184019087 - 184167628 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108653.1 NP_001102123.1 8 - 9 2962 68
XM_006237703.2 XP_006237765.1 9 - 10 3246 68

View Full Product Details