Assay Design



Gene Transcripts

Gene Symbol
Tom1l1
Entrez Gene ID
287622
Gene Name
target of myb1 (chicken)-like 1
Gene Aliases
RGD1562626, Srcasm
Chromosome Location
Chr.10: 78173847 - 78219805 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253859.1 NP_001240788.1 1 - 2 60 122
GenBank mRNA BC169112.1 - 1 - 2 58 122
CK596986.1 - 1 - 2 73 122

View Full Product Details