Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105844.1 | NP_001099314.1 | 2 - 3 | 601 | 85 |
XM_008768263.1 | XP_008766485.1 | 3 - 4 | 439 | 85 | |
XM_008768264.1 | XP_008766486.1 | 3 - 4 | 324 | 85 | |
XM_008768265.1 | XP_008766487.1 | 3 - 4 | 467 | 85 |