Assay Design



Gene Transcripts

Gene Symbol
Tmigd1
Entrez Gene ID
363654
Gene Name
transmembrane and immunoglobulin domain containing 1
Gene Aliases
RGD1307444, Tmigd
Chromosome Location
Chr.10: 63262554 - 63274598 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135029.1 NP_001128501.1 3 - 4 661 66

View Full Product Details