Assay Design



Gene Transcripts

Gene Symbol
Tusc5
Entrez Gene ID
360576
Gene Name
tumor suppressor candidate 5
Gene Aliases
DSPB1
Chromosome Location
Chr.10: 63471330 - 63491713 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039163.1 NP_001034252.1 1 - 2 830 110
GenBank mRNA AB218813.1 - 1 - 2 830 110

View Full Product Details