Assay Design



Gene Transcripts

Gene Symbol
Wfikkn1
Entrez Gene ID
363555
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
Gene Aliases
Oc29, RGD1565835
Chromosome Location
Chr.10: 15243429 - 15245757 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129776.1 NP_001123248.1 1 - 2 179 66

View Full Product Details