Assay Design
Gene Transcripts
- Gene Symbol
- Cenpv
- Entrez Gene ID
-
501702
- Gene Name
- centromere protein V
- Gene Aliases
-
Prr6, RGD1565681
- Chromosome Location
-
Chr.10: 48846513 - 48861895 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_577104.6 |
XP_577104.3 |
3 - 4 |
1464 |
84 |
View Full Product Details