Assay Design



Gene Transcripts

Gene Symbol
Hes7
Entrez Gene ID
287423
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
-
Chromosome Location
Chr.10: 55707164 - 55710518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105792.1 NP_001099262.1 1 - 2 48 65
XM_008767794.1 XP_008766016.1 1 - 2 43 65

View Full Product Details