Assay Design



Gene Transcripts

Gene Symbol
Ccdc78
Entrez Gene ID
302996
Gene Name
coiled-coil domain containing 78
Gene Aliases
RGD1560244
Chromosome Location
Chr.10: 15156074 - 15160039 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767616.1 XP_008765838.1 3 - 4 415 115
GenBank mRNA BC168182.1 - 3 - 4 347 115

View Full Product Details