Assay Design



Gene Transcripts

Gene Symbol
Gm2a
Entrez Gene ID
282838
Gene Name
GM2 ganglioside activator
Gene Aliases
-
Chromosome Location
Chr.10: 40438394 - 40450927 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172335.2 NP_758838.2 2 - 3 303 64
GenBank mRNA AB051391.1 - 2 - 3 262 64
BC072474.1 - 2 - 3 303 64

View Full Product Details