Assay Design



Gene Transcripts

Gene Symbol
Gm2a
Entrez Gene ID
282838
Gene Name
GM2 ganglioside activator
Gene Aliases
-
Chromosome Location
Chr.10: 40438394 - 40450927 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172335.2 NP_758838.2 3 - 4 483 85
GenBank mRNA AB051391.1 - 3 - 4 442 85
BC072474.1 - 3 - 4 483 85

View Full Product Details