Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109031.1 | NP_001102501.1 | 3 - 4 | 278 | 103 |
XM_006246393.2 | XP_006246455.1 | 4 - 5 | 384 | 103 | |
XM_006246394.2 | XP_006246456.1 | 3 - 4 | 499 | 103 | |
XM_006246395.2 | XP_006246457.1 | 3 - 4 | 497 | 103 | |
XM_008767703.1 | XP_008765925.1 | 3 - 4 | 608 | 103 |