Assay Design



Gene Transcripts

Gene Symbol
Ccdc69
Entrez Gene ID
497906
Gene Name
coiled-coil domain containing 69
Gene Aliases
RGD1562251
Chromosome Location
Chr.10: 40392459 - 40419275 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109031.1 NP_001102501.1 4 - 5 381 106
XM_006246393.2 XP_006246455.1 5 - 6 487 106
XM_006246394.2 XP_006246456.1 4 - 5 602 106
XM_006246395.2 XP_006246457.1 4 - 5 600 106
XM_008767703.1 XP_008765925.1 4 - 5 711 106

View Full Product Details