Assay Design



Gene Transcripts

Gene Symbol
Ccdc69
Entrez Gene ID
497906
Gene Name
coiled-coil domain containing 69
Gene Aliases
RGD1562251
Chromosome Location
Chr.10: 40392459 - 40419275 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109031.1 NP_001102501.1 6 - 7 600 80
XM_006246393.2 XP_006246455.1 7 - 8 706 80
XM_008767703.1 XP_008765925.1 6 - 7 930 80

View Full Product Details