Assay Design



Gene Transcripts

Gene Symbol
Gosr2
Entrez Gene ID
64154
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
-
Chromosome Location
Chr.10: 91736779 - 91756081 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031685.2 NP_113873.1 3 - 4 208 85
GenBank mRNA AF007549.1 - 3 - 4 222 85
BC061994.1 - 3 - 4 208 85
U91539.1 - 3 - 4 207 85

View Full Product Details