Assay Design



Gene Transcripts

Gene Symbol
Rilp
Entrez Gene ID
287531
Gene Name
Rab interacting lysosomal protein
Gene Aliases
-
Chromosome Location
Chr.10: 63659030 - 63662188 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105811.1 NP_001099281.1 6 - 7 924 153

View Full Product Details