Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053617.2 | NP_446069.1 | 2 - 3 | 184 | 121 |
GenBank mRNA | BC091133.1 | - | 2 - 3 | 148 | 121 |
BC107447.1 | - | 2 - 3 | 177 | 121 | |
DY312674.1 | - | 2 - 3 | 184 | 121 | |
FQ218610.1 | - | 2 - 3 | 189 | 121 | |
FQ218700.1 | - | 2 - 3 | 189 | 121 | |
FQ219645.1 | - | 2 - 3 | 256 | 121 |