Assay Design



Gene Transcripts

Gene Symbol
Cpb2
Entrez Gene ID
113936
Gene Name
carboxypeptidase B2 (plasma)
Gene Aliases
-
Chromosome Location
Chr.15: 57290849 - 57339762 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053617.2 NP_446069.1 4 - 5 410 91
GenBank mRNA BC091133.1 - 4 - 5 374 91
BC107447.1 - 4 - 5 403 91
DY312674.1 - 4 - 5 410 91
FQ218610.1 - 4 - 5 415 91
FQ218635.1 - 4 - 5 364 91
FQ218700.1 - 4 - 5 415 91
FQ219505.1 - 2 - 3 91
FQ219645.1 - 4 - 5 482 91

View Full Product Details