Assay Design



Gene Transcripts

Gene Symbol
Wbp4
Entrez Gene ID
114765
Gene Name
WW domain binding protein 4
Gene Aliases
Fbp21
Chromosome Location
Chr.15: 61745989 - 61772516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053766.1 NP_446218.1 8 - 9 854 151
XM_008770840.1 XP_008769062.1 5 - 6 1370 151
XM_008770841.1 XP_008769063.1 6 - 7 873 151
GenBank mRNA BC089052.1 - 8 - 9 854 151
FQ226684.1 - 7 - 8 636 151

View Full Product Details