Assay Design



Gene Transcripts

Gene Symbol
Rph3al
Entrez Gene ID
171123
Gene Name
rabphilin 3A-like (without C2 domains)
Gene Aliases
Noc2
Chromosome Location
Chr.10: 63989556 - 64140709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767941.1 XP_008766163.1 1 - 2 444 86
XM_008767943.1 XP_008766165.1 1 - 2 444 86
XM_008767944.1 XP_008766166.1 1 - 2 444 86

View Full Product Details