Assay Design
Gene Transcripts
- Gene Symbol
- Gucy2e
- Entrez Gene ID
-
79222
- Gene Name
- guanylate cyclase 2E
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 55835695 - 55851235 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024380.1 |
NP_077356.1 |
4 - 5 |
1449 |
92 |
GenBank mRNA |
L36029.1 |
- |
4 - 5 |
1449 |
92 |
View Full Product Details