Assay Design



Gene Transcripts

Gene Symbol
Slc26a11
Entrez Gene ID
360670
Gene Name
solute carrier family 26 (anion exchanger), member 11
Gene Aliases
-
Chromosome Location
Chr.10: 108479112 - 108504188 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247940.2 XP_006248002.1 8 - 9 1103 80

View Full Product Details