Assay Design



Gene Transcripts

Gene Symbol
Tekt3
Entrez Gene ID
287392
Gene Name
tektin 3
Gene Aliases
-
Chromosome Location
Chr.10: 49472520 - 49506473 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024739.1 NP_001019910.1 5 - 6 1019 81
GenBank mRNA BC097398.1 - 5 - 6 1019 81

View Full Product Details