Assay Design



Gene Transcripts

Gene Symbol
Ypel2
Entrez Gene ID
360590
Gene Name
yippee-like 2 (Drosophila)
Gene Aliases
RGD1311564
Chromosome Location
Chr.10: 74241032 - 74298599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108286.1 NP_001101756.1 3 - 4 509 119
XM_006247097.1 XP_006247159.1 3 - 4 494 119

View Full Product Details