Assay Design



Gene Transcripts

Gene Symbol
Nog
Entrez Gene ID
25495
Gene Name
noggin
Gene Aliases
-
Chromosome Location
Chr.10: 76811759 - 76813386 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012990.1 NP_037122.1 1 - 1 1339 138
GenBank mRNA AA859752.1 - 1 - 1 138
BF549968.1 - 1 - 1 362 138

View Full Product Details