Assay Design



Gene Transcripts

Gene Symbol
Hoxb2
Entrez Gene ID
100361765
Gene Name
homeobox B2
Gene Aliases
Hoxbes2
Chromosome Location
Chr.10: 84200859 - 84203065 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002727806.4 XP_002727852.1 1 - 2 413 138

View Full Product Details