Assay Design



Gene Transcripts

Gene Symbol
Fbxo39
Entrez Gene ID
303287
Gene Name
F-box protein 39
Gene Aliases
-
Chromosome Location
Chr.10: 58874021 - 58878445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246729.2 XP_006246791.1 1 - 2 87 82
GenBank mRNA BC082089.1 - 1 - 2 40 82

View Full Product Details