Assay Design



Gene Transcripts

Gene Symbol
Slitrk6
Entrez Gene ID
290467
Gene Name
SLIT and NTRK-like family, member 6
Gene Aliases
-
Chromosome Location
Chr.15: 95507632 - 95514259 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106057.1 NP_001099527.1 1 - 2 523 71

View Full Product Details