Assay Design



Gene Transcripts

Gene Symbol
Ubac2
Entrez Gene ID
361094
Gene Name
UBA domain containing 2
Gene Aliases
Phgdhl1
Chromosome Location
Chr.15: 108286379 - 108433531 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034937.2 NP_001030109.2 7 - 8 929 106
XM_006252509.2 XP_006252571.1 6 - 7 955 106
GenBank mRNA BC105842.1 - 2 - 3 312 106

View Full Product Details