Assay Design



Gene Transcripts

Gene Symbol
Slc39a14
Entrez Gene ID
306009
Gene Name
solute carrier family 39 (zinc transporter), member 14
Gene Aliases
-
Chromosome Location
Chr.15: 51982872 - 52029841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107275.1 NP_001100745.1 7 - 8 1160 78
XM_006252270.2 XP_006252332.1 7 - 8 1185 78

View Full Product Details